Black Qualitative Filter Paper, Grade 8613, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom 8613 has a smooth black finish designed for use in applications requiring visual examination of particles.
Specifications
- Catalog No:
- 47746-930
- 28342-052
- 28342-020
- 28320-555
- 47745-876
- 47745-878
- 28342-010
- 28342-012
- 47745-870
- 47745-872
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm
- 4 µm