Qualitative Filter Paper, Grade 631, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom Grade 631 is used for filtration of coarse particles and some gelatinous precipitates, air monitoring when high flow rate is needed, and in routine cleanup analysis of insoluble liquids.
This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.
Spécifications
- Catalog No:
- 47749-706
- 47749-686
- 47749-702
- 47749-704
- 47749-698
- 47749-700
- 47749-694
- 47749-696
- 47749-692
- 47749-708
- 47749-688
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- Retention:
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm