To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Cell Format:Cryopreserved
- Description:Tumor
- Size:0.1 mg
- Storage conditions:Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
- Storage buffer:Spleen Tumor lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
- Concentration:2 mg/mL
- Tested applications:WB
- Cat. no.:10062-626
- Source:Human
- Tissue:Spleen
- Supplier No.:1339
Spécifications
A propos de cet article
Spleen tumor tissue lysate was prepared by homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)
Tissue and cell debris was removed by centrifugation. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.