Glass Microfiber Filter Paper, Grade 131, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Grade 131 is a binder-free microfiber glass filter paper.
Spécifications
- Catalog No:
- 47749-560
- 47745-796
- 47749-556
- 47742-552
- 47749-558
- 47749-552
- 47749-554
- 47749-548
- 47749-550
- 47745-798
- 38010-685
- 38010-670
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Retention:
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm