Glass Microfiber Filter Paper, Grade 111, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Grade 111 is a binder-free microfiber glass filter paper that offers fine particle retention with high filtration speed and good loading capacity.
Spécifications
- Catalog No:
- 47745-772
- 47749-530
- 47745-786
- 47745-788
- 47745-782
- 47749-528
- 47745-784
- 47749-526
- 47745-778
- 47745-780
- 47745-774
- 47745-776
- 38010-100
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Retention:
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm