Order Entry
ContactUsLinkComponent
Human Recombinant IL33 (from E. coli)
Human Recombinant IL33 (from E. coli)
# de catalogue 10072-014
Fournisseur:  Prosci
Numéro CAS:  
IL-33 Recombinant Protein Species reactivity: Human Host: E. coli alternate name: NF-HEV
Human Recombinant IL33 (from E. coli)
# de catalogue 10072-014
Fournisseur:  Prosci
Numéro CAS:  

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein synonyms:
    NFEHEV|C9orf26|IL1F11|NF-HEV|IL-33|IL-33 DVS27|NFHEV|Interleukin-1 family member 11|Interleukin-33
  • Protein/peptide name:
    IL33
  • Purity:
    >95%
  • Molecular weight:
    22 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested applications:
    ELISA, Western Blot
  • Cat. no.:
    10072-014
  • Supplier No.:
    95-111

Spécifications

A propos de cet article

Interleukin-33 (IL-33) is a recently identified member of the IL-1 family of cytokines whose other members include IL-1, IL-1Ra and IL-18 (1,2). Its receptor has been shown to be ST2, an IL-1 receptor family member that also acts as a negative regulator of TLR-IL-1R signaling (1,3) and IL-1R accessory protein (IL-1RAcP) (4). Receptor binding of IL-33 activates NF-B and MAP kinases and induces the expression of TH2-associated cytokines such as IL-4, IL-5 and IL-6. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine (2). Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties (5). This recombinant protein represents the cleaved and presumably activated form of IL-33, but has not been tested in any biological assays.

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

100mM sodium phosphate, 500mM NaCl, 25 mM imidazole, 10% gycerol

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought