Order Entry
ContactUsLinkComponent
Human Recombinant TSG (from E. coli)
Human Recombinant TSG (from E. coli)
# de catalogue 10072-756
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant TSG (from E. coli)
# de catalogue 10072-756
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    Biological activity was determined by its ability to neutralize BMP - 6 induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED50 for this effect is < 2.0 μg/mL of TSG.
  • Protein synonyms:
    TSG|PSEC0250|Twisted gastrulation protein homolog 1
  • Protein/peptide name:
    TSG
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-756
  • Supplier No.:
    40-205

Spécifications

A propos de cet article

Twisted Gastrulation Protein (TSG) is a secreted BMP binding protein structurally related to the BMP antagonists Chordin and Noggin. TSG can inhibit BMP activity by binding directly to BMP proteins, and can act either as a BMP4 agonist or antagonist (depending on the specific biochemical environment) by binding to the BMP4/Chordin complex. Recombinant human TSG is a 199 amino acid 22.2 kDa protein containing the BMP/TGFβ binding portion of the full length TSG protein.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought