Order Entry
ContactUsLinkComponent
Human Recombinant BMP6 (from HEK293 cells)
Human Recombinant BMP6 (from HEK293 cells)
# de catalogue 10072-754
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP6 (from HEK293 cells)
# de catalogue 10072-754
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.002 mg
  • Biological activity:
    Biological activity was determined by its ability to induce alkaline phosphatase production by ATDC - 5 cells. The expected ED50this effect is 0.03 - 0.06 μg/mL.
  • Protein synonyms:
    VGR1|Bone morphogenetic protein 6|VGR|BMP-6|VG-1-related protein
  • Protein/peptide name:
    BMP6
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-754
  • Supplier No.:
    40-204

Spécifications

A propos de cet article

TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling and maintenance of a variety of other tissues and organs. Increasing evidence indicates that BMP-Smad signaling has a tumor suppressing activity and that BMPs can inhibit tumor growth. BMP-6 is abnormally expressed in breast cancer cell lines; however, its function in promoting breast cancer development is unknown. Recombinant human BMP-6 is a 26.2 kDa homodimeric glycoprotein consisting of two 117 amino acid subunits, which correspond to amino acid residues 397 to 513 of the full-length BMP-6 precursor.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought