Order Entry
ContactUsLinkComponent
Human Recombinant Cdmp-2 (from E. coli)
Human Recombinant Cdmp-2 (from E. coli)
# de catalogue 10072-752
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Cdmp-2 (from E. coli)
# de catalogue 10072-752
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    The ED50 was determined by its ability to induce alkaline phosphatase production by ATDC - 5 chondrogenic cells in the range of 2.0 - .0 μg/mL.
  • Protein synonyms:
    SGM1|CDMP2|KFS1|MCOP4|SCDO4|MCOPCB6|LCA17|BMP13|GDF16GDF-6|KFSL|KFM|KFS
  • Protein/peptide name:
    Cdmp-2
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-752
  • Supplier No.:
    40-202

Spécifications

A propos de cet article

BMP-13 is expressed in hypertrophic chondrocytes during embryonic development of long bones. Continued postnatal expression of BMP-13 in articular cartilage suggests that it plays a regulatory role in the growth and maintenance of articular cartilage. Adenovirus-mediated BMP-13 gene transfer to rabbit bone marrow stem cells have been reported to augment periosteal repair of osteochondral defects. The functional form of BMP-13/CDMP-2 is a disulfide-linked homodimer of two 120 amino acid polypeptide chains. This 27.5 kDa protein is obtained by proteolytic processing of a biologically inactive precursor protein of 97.7 kDa. Recombinant human BMP-13/CDMP-2 is a 27.0kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought