Vikan® Telescopic Handles, Aluminum, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This extendable handle is often used with the pad holder and deck scrubs for reaching high walls and tanks.
- Aluminum and polypropylene materials
Also used with CIP brushes for cleaning on top of overhead pipes.
Width: 1.25"
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought