Vikan® Fiberglass Handles, 51", Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This standard broom handle can be used with all brooms, squeegees and scrapers.
Fiberglass and polypropylene materials
The handle with the attached item should reach to the chin of the user to prevent occupational back injuries.
Width: 1.25"
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought