Vikan® Aluminium Handles, 51", Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This standard broom handle can be used with all brooms, squeegees and scrapers.
- Aluminum and polypropylene materials
The handle with the attached item should reach to the chin of the user to prevent occupational back injuries.
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought