CPEP Fusion, Physics of Fundamental Energy Charts
# de catalogue 470122-544
Fournisseur: CONTEMPORARY PHYSICS EDUCATION
Spécifications
- Description:Laminated Placemat, 16 x 11"
- Cat. no.:470122-544
- Supplier No.:397341
Spécifications
A propos de cet article
Visualize fusion and energy.
- Students will be able to compare the energy released in a variety of conversion processes, including combustion, fission and fusion
Developed by the Fusion and Plasmas Group of the Contemporary Physics Education Project (CPEP), Fusion presents up-to-date information on plasma physics and fusion reactions, examples of solar and terrestrial fusion reactions, explains how Einstein’s famous E=mc2 describes the process of fusion and offers illustrations of everyday and exotic plasmas.
Customers Who Bought This Also Bought