Plastic Coated Solvent Delivery Bottles, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Solvent reservoir for use in HPLC mobile phase delivery.
Outer joint at the top on all sizes is polished, thereby making an excellent seal between the PTFE cap and the bottle. Optional stopper permits convenient storage of solvent.
Customers Who Bought This Also Bought