DiamondⓇ White Glass Microscope Slides, Plain, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
Globe scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
- Produced from the finest quality optical white glass
- Dimensions: 25×75 mm
- Thickness: 1 mm
DiamondⓇ White Glass plain microscope slides are available with 90° or clipped corners. The slides are pre-cleaned and supplied ready for use.
Packaging: Slides are packaged 72 per box, 20 boxes per case (10 gross).
Customers Who Bought This Also Bought