Precision™ Compact Ovens, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Conserve valuable benchtop space with Precision™ Compact Ovens, featuring mechanical or gravity convection.
Ideal for drying and baking applications. Ovens feature a double-wall interior with 2.5 cm (1") silica-based insulation and powder-coated cold-rolled steel exterior. Pilot light visually indicates oven operation status.
Certifications: UL/cUL listed.
Spécifications
- Catalog No:
- 97018-372
- 97018-376
- Temperature range:
- Ambient +5° to 210°C
- Ambient +5° to 210°C
- Volume:
- 1.7 cu. ft.
- 1.7 cu. ft.
- Int. depth:
- 29 cm (11.5")
- 29 cm (11.5")
- Int. height:
- 41 cm (16.2")
- 41 cm (16.2")
- Int. width:
- 41 cm (16")
- 41 cm (16")