To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:tubulin, beta 3 class III
- Clonality:Polyclonal
- Clone:5G8
- Gene ID:Q13509
- Host:Mouse
- Reactivity:Human,Mouse
- Antigen symbol:TUBB3
- Conjugation:Unconjugated
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TUBB3
- Storage buffer:PBS Buffer containing 0.02% sodium azide.
- Storage temperature:2-8 °C for one week and –20 °C as an undiluted liquid for up to two years.
- Shipping temperature:4 °C
- Immunogen:C-terminal peptide of beta III tubulin.
- Cat. no.:10068-198
- Supplier No.:XG-6195
Spécifications
A propos de cet article
This antibody can be used at 1:1000 to 1:4,000 for western blot, ELISA testing, and IHC/ICC.
PBS Buffer containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse