To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:fat mass and obesity associated
- Clonality:Monoclonal
- Clone:5-2H10
- Gene ID:Q9C0B1
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:FTO
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FTO
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:58
- Shipping temperature:4 °C
- Immunogen:Synthetic peptide from the N-terminal region of rat Fto.
- Purification:Protein G purified
- Cat. no.:10075-604
- Supplier No.:50-122
Spécifications
A propos de cet article
Protein G purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: FTO
Clonality: Monoclonal
Clone: 5-2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat