To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:breast cancer 1, early onset
- Clonality:Polyclonal
- Gene ID:P38398
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:BRCA1
- Conjugation:Unconjugated
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:BRCA1
- Storage buffer:PBS containing 0.02% sodium azide.
- Storage temperature:Cyclin E1 antibody can be stored at 4 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:Peptide corresponding to amino acids 43-52 of cyclin E1.
- Purification:Affinity-purified
- Cat. no.:10070-998
- Supplier No.:47-111
Spécifications
A propos de cet article
Affinity-purified
PBS containing 0.02% sodium azide.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human