To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ubiquitin-conjugating enzyme E2S
- Clonality:Polyclonal
- Gene ID:Q16763
- Host:Goat
- Reactivity:Human
- Antigen symbol:UBE2S
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:UBE2S
- Storage buffer:E2-EPF antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:E2-EPF antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of E2-EPF.
- Purification:E2-EPF antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10112-520
- Supplier No.:45-513
Spécifications
A propos de cet article
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 35-40 kDa band observed in human liver lysate at 1 ?g of primary, this band was successfully blocked by incubation with the immunizing peptide. Please note that the band we observe is different from the predicted size of approximately 25 kDa according to NP_055316.
E2-EPF antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: UBE2S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: