To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:bone morphogenetic protein 7
- Clonality:Monoclonal
- Clone:M123-11A
- Gene ID:P18075
- Host:Mouse
- Reactivity:Human
- Antigen symbol:BMP7
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.5 mg
- Form:Lyophilized
- Antigen synonyms:BMP7
- Shipping temperature:RT
- Immunogen:BMP-7 antibody is produced in mice using highly pure (>98%) recombinant human BMP-7 as the immunizing antigen.
- Purification:This IgG1K antibody was purified from cell culture by Protein G affinity chromatography.
- Cat. no.:10097-920
- Supplier No.:38-232
Spécifications
A propos de cet article
BMPs (bone morphogenetic proteins) belong to the TGF beta superfamily of structurally related signaling proteins. Members of this superfamily are widely represented throughout the animal kingdom and have been implicated in a variety of developmental processes. Proteins of the TGF beta superfamily are disulfide-linked dimers composed of two 12-15 kDa polypeptide chains. As implied by their name, BMPs initiate, promote and regulate bone development, growth, remodeling and repair.
ELISA: In a sandwich ELISA (assuming 100 µL/well), when used as a capture antibody at a concentration of 2.0-4.0 µg/mL, this antibody will detect at least 100 pg/mL of recombinant human BMP-7/OP-1 when used with our biotinylated antigen affinity purified-hBMP-7 as the detection antibody at a concentration of approximately 0.5-1.0 µg/mL.
This IgG1K antibody was purified from cell culture by Protein G affinity chromatography.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: BMP7
Clonality: Monoclonal
Clone: M123-11A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human