To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:kelch-like family member 5
- Clonality:Polyclonal
- Gene ID:Q96PQ7
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KLHL5
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:KLHL5
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:79 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store KLHL5 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human KLHL5.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10102-122
- Supplier No.:25-485
Spécifications
A propos de cet article
The kelch-repeat protein family is a recently found new kind of actin-binding protein. It is characterized by tandemly arranged motifs of about 50 amino acids. Most members of the kelch-repeat family were cytoskeletal proteins implicated in various cellular processes, such as actin cytoskeleton interaction, cytoplasmic sequestration of transcription factors and cell morphology. During the large-scale sequencing analysis of a human fetal brain cDNA library we found a novel kelch-like protein gene 5, KLHL5, KLHL5 has high identity with Drosophila kelch protein and many other family members. A novel splicing variants of KLHL5, named KLHL5b and the expression pattern of KLHL5b in many tissues.
KLHL5 antibody can be used for detection of KLHL5 by ELISA at 1:1562500. KLHL5 antibody can be used for detection of KLHL5 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KLHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat