Dissolved Oxygen Probes, IDS FDO®, for Multi 3410 / 3420 / 3430, WTW
Fournisseur: WTW
Certificats
A propos de cet article
Optical dissolved oxygen probes for field and laboratory applications.
- Extremely fast and precise
- Factory calibrated cap with built-in data chip
- Proven GLT (Green Light Technology) probe for long operation life
- Slender design (suitable for Winkler bottles)
- Bevelled membrane repells interference that can be caused by air bubbles
Ordering information: Various cable lengths available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.