A propos de cet article
Waterproof handheld meters designed for unlimited outside use.
- Large display allowing excellent readability, even under bad lighting conditions
- Robust housing and sealed silicone keypad with protection class IP 66/67
- Up to 2500 hour continuous operation time with one battery set
For challenging measurements in the field and plant, e.g
The Oxi 3205 is a small version of the Oxi 3210 without memory but including simplified functions.
For professional field measurements requiring documentation and data transfer. The Oxi 3310 IDS models enable digital measuring of optical DO. The optical DO sensor FDO® 925 is not only recommended for BOD measuring, but also for field and process applications.
Same specifications as Oxi 3210, with:
Extended memory for 5000 GLP-compliant data
Time-controlled datalogger with adjustable intervals for long-term monitoring
Waterproof mini USB interface to a PC.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 97043-136
- 97043-118
- 97043-120
- 97043-134
- Model:
- Oxi 3310
- Oxi 3205
- Oxi 3205
- Oxi 3310
- DO concentration:
- 0.00 - 19.99 / 0 - 90 mg/L
- 0.00 - 19.99 / 0 - 90 mg/L
- 0.00 - 19.99 / 0 - 90 mg/L
- 0.00 - 19.99 / 0 - 90 mg/L
- DO saturation:
- 0.0 - 199.9 % / 0 - 600 %
- 0.0 - 199.9 % / 0 - 600 %
- 0.0 - 199.9 % / 0 - 600 %
- 0.0 - 199.9 % / 0 - 600 %
- DO barometric pressure range:
- 0 - 199.9 / 0 - 1250 mmHg
- 0 - 199.9 / 0 - 1250 mmHg
- 0 - 199.9 / 0 - 1250 mmHg
- 0 - 199.9 / 0 - 1250 mmHg
- Temperature range:
- –5.0…+50.0 °C
- –5.0…+50.0 °C
- –5.0…+50.0 °C
- –5.0…+50.0 °C
- Temperature accuracy:
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- Compliance:
- IP 66/67, GLP
- IP 66/67
- IP 66/67
- IP 66/67, GLP