A propos de cet article
Rugged lines of conductivity meters with a range of features, benefits, and price points to meet every application and budget!
- Rugged, waterproof housing is great for the field or the lab
- Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad
- Silicone rubber keys provide tactile feedback for error-free operation
Cond 3110 - Designed for users who desire a simple-to-use, yet dependable instrument. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.
Cond 3310 - Innovative waterproof USB connection for ultra-flexible data output. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results. Meter provide extended data memory for logging up to 5000 GLP-compliant data sets. Time-controlled datalogging function offers adjustable data collection intervals—ideal for long-term field monitoring. A special “sleep” mode saves power and extends battery life when performing long-term monitoring. Nonvolatile memory saves all stored data even if battery power is lost. Data transfer to PC is made simple via the unique waterproof mini-USB interface.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 97043-140
- 97043-156
- 97043-158
- Model:
- Cond 3110
- Cond 3310
- Cond 3310
- Conductivity range:
- 0 - 1000 mS/cm
- 0.0 - 199.9 / 200 - 1999 (µS/cm); 2.00 - 19.99 / 20.0 - 199.9 / 200 - 1000 (mS/cm)
- 0.0 - 199.9 / 200 - 1999 (µS/cm); 2.00 - 19.99 / 20.0 - 199.9 / 200 - 1000 (mS/cm)
- Conductivity accuracy:
- ±0.5%
- ±0.1 / ±1 (µS/cm); ±0.01 / ±0.1 / ±1 (mS/cm)
- ±0.1 / ±1 (µS/cm); ±0.01 / ±0.1 / ±1 (mS/cm)
- Conductivity cell constant:
- Fixed: 0.475 cm⁻¹ / 1.0 cm⁻¹
Calibratable: 0.450 - 0.500 cm⁻¹ / 0.585 - 715 cm⁻¹ / 0.800 - 1.200 cm⁻¹ - Fixed: 0.475 cm⁻¹ / 0.01 cm⁻¹
Calibrated: 0.450-0.500 cm⁻¹ / 0.585-0.715 cm⁻¹ / 0.800-1.200 cm⁻¹
Adjustable: 0.090-0.110 cm⁻¹
Standard: 0.01 mol/l KCl - Fixed: 0.475 cm⁻¹ / 0.01 cm⁻¹
Calibrated: 0.450-0.500 cm⁻¹ / 0.585-0.715 cm⁻¹ / 0.800-1.200 cm⁻¹
Adjustable: 0.090-0.110 cm⁻¹
Standard: 0.01 mol/l KCl
- Temperature range:
- –5.0…+105.0 °C
- –5.0…+105.0 °C
- –5.0…+105.0 °C
- Temperature accuracy:
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- Compliance:
- IP 66/67
- IP 66/67, GLP
- IP 66/67, GLP