Whatman™ Grade 802 Qualitative Filter Papers, Whatman products (Cytiva)
Fournisseur: Cytiva
Certificats
A propos de cet article
A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
- Highest retention among standard filters
- Slow flow rate
- Fine-particle retention
- For qualitative analysis
The filter is wet-strengthened and for normal qualitative application it will not introduce any significant impurities into the filtrate. However, it is not recommended for Kjeldahl nitrogen analysis.
Customers Who Bought This Also Bought