A propos de cet article
CTC PAL autosamplers are one of the most popular sampling devices on the market today
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. S-Line syringes deliver great performance in everyday use for GC injections at an exceptional value. The Gastight® version of the S-Line syringe is an ideal choice for GC sample injections. It incorporates a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit in the glass barrel, the tip wipes the interior of the syringe barrel free of sample, minimizing sample carryover. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Customers rely on their ability to sample from many different vial and well types and on their ultimate platform flexibility. The syringe is the heart of every injection that the GC PAL system performs. The latest generation of Hamilton autosampler syringes is recommended by CTC for use in the popular GC PAL systems: PAL HTC = xt, PAL HTS = xt, PAL HTX = xt.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 89498-960
- 89498-962
- 89498-964
- 89498-966
- 89498-968
- 89498-970
- Application:
- GC
- GC
- GC
- GC
- GC
- GC
- Autoclavable:
- No
- No
- No
- No
- No
- No
- Needle length:
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- Point style:
- AS
- AS
- AS
- AS
- AS
- AS