Dura-Clamp, Dynalon
# de catalogue 62849-112
Fournisseur: Dynalon
Spécifications
- Cat. no.:62849-112
- L×W×H:1 x 1/3 x 2/3"
- Supplier No.:670715
Spécifications
A propos de cet article
Polypropylene Dura-Clamp tubing flow valve is easily operated with one hand and will not rust or corrode
Accepts tubing O.D. up to 1/4".
Customers Who Bought This Also Bought