Borosilicate Glass Culture Tubes, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
These tubes are made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from soda lime glass tubes and are used for tissue culture and blood banks.
- Excellent chemical resistance and durability
- Features well-rounded bottoms and smooth fire-polished rims
TubeSaver™ packaging system helps prevent breakage and contamination during shipping and storage. Dimensional specifications are rigidly controlled for reliable, uniform results.
Please note: The capacity given below indicates the brim capacity (overflow capacity).
Certifications: An ISO 9001:2008 certified factory product.
Packaging: The tubes are neatly stacked in heavy-duty, shrink wrapped boxes.
Frequently Bought Together