ContactUsLinkComponent
Cotton Tipped Applicator Swab, Dynalon
Cotton Tipped Applicator Swab, Dynalon
# de catalogue 30621-018
Fournisseur:  Dynalon
Swab Cottn Reg Wood 6In Cs1000
Cotton Tipped Applicator Swab, Dynalon
# de catalogue 30621-018
Fournisseur:  Dynalon
Spécifications
  • Length:
    6"
  • Material:
    Wood
  • Cat. no.:
    30621-018
  • Supplier No.:
    394365
Spécifications
A propos de cet article
Disposable cotton swabs manufactured of USP grade purified cotton firmly attached to one end of wood shaft.
May be autoclaved. Useful in bacteriology labs for inoculating broths, plated media or for obtaining culture specimens. Also useful for cleaning delicate instruments.
Customers Who Bought This Also Bought