Enviro-Swab, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Reticulated design is precision molded from superior grade, open cell polyurethane 100 ppi foam, consistently demonstrating low particulation and chemical cleanliness
Enviro-Swabs™ are non-silicone formulated, hot-wire cut and slit (not die cut and sealed). A thermally bonded head to handle eliminates the use of adhesives for maximum reliability and performance. These swabs are non-linting and non-scratching. They may be used with alcohol, acetone, oils, greases and solvents.
Customers Who Bought This Also Bought