Joint Sleeve, PTFE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Unique manufacturing process permits superior quality PTFE joint sleeves
Elongated cones of micro-thin virgin PTFE film tapered to fit snugly around glass joints. Approximately 0.05 mm (0.002") thick with taper of approximately 1.10 on diameter. Eliminates the need for lubrication and frozen, leaky ground joints. Can be used over a temperature range of -75° to +260° C.
Customers Who Bought This Also Bought