Angle Cut Broom, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
The angled design of this broom makes it easy to reach into narrow spaces between equipment when sweeping debris before the scrubbing action starts
It is particularly effective for collecting large particles of debris like meat and vegetables in a moist environment. Block material is polypropylene; bristle material is PET.
Certifications: FDA Compliant Raw Material.
Customers Who Bought This Also Bought