Biohazard Disposal Bags
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
Biohazard disposal bags are ideal for processing biohazardous materials that require autoclaving prior to disposal.
- Bags are designed for personal or lab use
- Available in sizes to suit all needs
- Autoclavable to 121 °C (250 °F)
The polypropylene bags include an indicator patch that darkens when autoclaved. Handy-size bag holder is ideal for collecting tips and tubes.
Customers Who Bought This Also Bought