Stirring Rod, Solid, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Pure PTFE solid stirring rod with flat tapered ends
Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.
Customers Who Bought This Also Bought