Stir Bar, Cross Head, PTFE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Designed for round or flat bottom labware and test tubes
Greater mixing action at bottom, side, and top of container, even at low stirring speeds. Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance and is autoclavable. Always store in pairs to maintain magnetic strength.
Customers Who Bought This Also Bought