Stir Bar, Extra Large, PTFE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Extra large polygonal stir bar for use in very large containers
Powerful permanent Alnico V magnetic core. PTFE has excellent chemical resistance and can be used up to 260°C. Always store in pairs to maintain magnetic strength.
Customers Who Bought This Also Bought