Stir Bar, Octagonal, PTFE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Octagonal stirring bars feature powerful permanent Alnico V magnetic cores
PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.
Customers Who Bought This Also Bought