Permafrost Economy Frosted Microscope Slides, Thermo Scientific
Fournisseur: Erie Scientific
Certificats
A propos de cet article
Permafrost Slides are designed for use in general laboratory applications, such as urinalysis, microbiology, hematology, and histology
All Permafrost slides are pre-cleaned, measure 25x75x1mm, feature smoothly ground edges, and are manufactured from white Swiss glass. They are packaged approximately 72 slides per pack, 20 packs per case.
Permafrost slides feature a ¾" frosted end available in 10 different colors.
Customers Who Bought This Also Bought