Nalgene® Biotainer Replacement Closures, Thermo Scientific
Catalog # 89429-586
Supplier: Thermo Fisher Scientific
Specifications
- Cap size:48 mm
- Description:Biotainer Replacement Closures
- Cat. no.:89429-586
- Supplier No.:362515-0480
Specifications
About this item
Polypropylene Biotainer Replacement Closures are designed to protect the contents within InVitro™ Biotainer™ bottles and carboys
Closures feature silicone liners and are compatable with 1L, 2L, 5L, 10L, and 20L polycarbonate Biotainers. Closure size is 48mm. Sterile.
Customers Who Bought This Also Bought