Blood Bank Cross Match Holders, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Blood bank cross match holders are manufactured of clear acrylic shelves with white bases and sides
For typing, screening and other blood bank applications. For blood donor processing. Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping and screening. No on rack labeling supplied.
Ordering information: Additional sizes and materials are available upon request.
Customers Who Bought This Also Bought