Thick Chromatography Paper, Grade 243, Ahlstrom
# de catalogue 47749-850
Fournisseur: Ahlstrom
Spécifications
- Dimensions:46 x 57 cm
- Absorption speed:180 mm / 30 min
- Thickness:0.95 mm
- Cat. no.:47749-850
- Supplier No.:2438-4657
Spécifications
A propos de cet article
Ahlstrom Grade 243 is one of the thicker chromatography papers available from Ahlstrom
It offers moderately high flow and absorption and is recommended for chromatography with heavy solute loading.
Customers Who Bought This Also Bought