Thick Chromatography Paper, Grade 237, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom Grade 237 is a medium thick 100% cotton chromatography paper recommended for general applications with medium-heavy solute loading
It is also ideal for use as wicks in protein or nucleic acid blotting.
Spécifications
- Catalog No:
- 47745-896
- 47745-892
- 47745-894
- 47745-890
- Thickness:
- 0.42 mm
- 0.42 mm
- 0.42 mm
- 0.42 mm
- Absorption speed:
- 150 mm / 30 min
- 150 mm / 30 min
- 150 mm / 30 min
- 150 mm / 30 min