Economy Bottle Carrier, Dynalon
# de catalogue 30621-004
Fournisseur: Dynalon
Spécifications
- Description:HDPE utility carrier for 6 bottles
- Cat. no.:30621-004
- L×W×H:13.56×10.12×11"
- Weight:0.65 lbs.
- Supplier No.:408175
Spécifications
A propos de cet article
Lightweight, sturdy bottle carrier made from High Density Polyethylene
Transports up to 6 bottles with a diameter of 3 1/2 inches or smaller (usually 250 - 1L sizes). The handle is approximately 6 inches in height.
Customers Who Bought This Also Bought