Spatula Scrapers, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
A short handled unit specially designed as a scraper and spatula
Scrape and clean any hard to clean containers. 13 1/2" overall length. Polyethylene blade is 4 1/2" wide, 7" long. Complete with large hand-grip.
Customers Who Bought This Also Bought