Drying Rack, High Impact Polystyrene, Dynalon
# de catalogue 30620-320
Fournisseur: Dynalon
Spécifications
- Length:177/10"
- Width:44/10"
- Description:Drying Rack
- Height:248/10"
- Cat. no.:30620-320
- Weight:5.7 lbs.
- Supplier No.:259184
Spécifications
A propos de cet article
This 72 place drying rack is molded of high impact polystyrene
It comes supplied with 72 large pegs, draining tubes, fastening stoppers and expansion hanging hooks which allow coupling with additional racks. The pegs are 4" long and the drip ledge measures 3". Expandable fasteners to connect additional racks included and interchangeable removable pegs
Customers Who Bought This Also Bought