Shorty TLC Developing Tank, Kimble Chase
Catalog # KT416180-1020
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:SHORTY TLC developing tank
- Cat. no.:KT416180-1020
- Supplier No.:416180-1020
Specifications
About this item
The standard 20 x 20 cm TLC plate tank is difficult to use with 10 x 10 cm plates, so the “SHORTY” tank was developed
Lids are available with holes drilled for #0 stoppers to allow addition of reagents without disturbing the equilibrium of the saturated environment.
It will accommodate 5 x 10 or 10 x 20 plates, turned long side down. Both have a centering baffle to support plates during development and are supplied with lids.
Customers Who Bought This Also Bought