Sample Cups, Polystyrene, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Clear polystyrene cups for use with older auto analyzers.
Also useful for general micro use. Conical bottom. 26mm height. LDPE Stopper available
Customers Who Bought This Also Bought