ContactUsLinkComponent
Plant Growth Chambers, Extra Large Capacity, Caron Products
Plant Growth Chambers, Extra Large Capacity, Caron Products
Fournisseur:  Caron Products
   Sustainable Options Available
Certificats
Plant Growth Chambers, Extra Large Capacity, Caron Products
Plant Growth Chambers, Extra Large Capacity, Caron Products
Fournisseur:  Caron Products
   Sustainable Options Available
Certificats
Spécifications
  • Catalog No:
  • 10068-342
  • 10068-344
  • 10068-370
  • 10068-346
  • 10068-356
  • 10068-358
  • 10068-360
  • 10068-308
  • 10068-310
  • 10068-364
  • 10068-336
  • 10068-362
  • 10068-338
  • 10068-368
  • 10068-340
  • 10068-366
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Energy Efficient
  • Temperature range:
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Lights On: 10°C to 50°C; Lights Off: 5°C to 50°C
  • Temperature sensor:
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • 3-wire, RTD
  • Temperature control:
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • ±0.2°C
  • Material:
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Shelf dimensions:
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
  • 55W x 67D cm D (21.6 x 26.5")
Spécifications
Informations connexes
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product.
PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
PM Service Kits for Accessories help to minimize downtime by providing routine maintenance items for accessories. See table below for more information.