Corning 384-well High Content Screening Microplates with Glass Bottom
Supplier: Corning
Certificates
About this item
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 µm is well suited for imaging microscopy
- Bottom flatness <50 µm to ensure planarity for imaging devices
- Low background fluorescence and minimal crosstalk provide the highest possible optical quality for cell-based assays
Specifications
- Catalog No:
- 89511-330
- 89511-322
- 30621-098
- 89511-326
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Bedford
- I-RECs, ISO 14001 - Bedford
- RECs, ISO 14001 - Kennebunk
- I-RECs, ISO 14001 - Bedford